Tisk…
Anti-IgG Goat Polyclonal Antibody (iFluor® 514)
Dodavatel: AAT BIOQUEST
50060 50061
AATB50060EA 3210 CZK
AATB50060 AATB50061
Anti-IgG Goat Polyclonal Antibody (iFluor® 514)
Protilátky
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.
  • Affinity purified
  • Easily reconstituted
Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).
Upozornení: For research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 514
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human