Cresyl violet acetate, pure, certified
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Cresyl echt violet, Cresyl violet, Cresyl fast violet, 5,9-Diaminobenzo[a]phenoxazin-7-ium acetate, 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt, Cresyl Violet acetate, Gentian violet
405760025
405760100
405760500
ACRO405760025EA
2970
CZK
ACRO405760025
ACRO405760100
ACRO405760500
Cresyl violet acetate, pure, certified
Cresyl violet (acetate)
Vzorec:
C₂₅H₃₀ClN₃ Bod tání: 140…143 °C Hustota: 1,19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013151 CAS číslo: 548-62-9 EINECS: 208-953-6 ADR: 9,III Merck Index: 13,04406 |
Výsledky specifikačního testu
Appearance | Dark green to black Crystalline powder |
Infrared spectrum | Conforms |
Dye content | ≥65 % |
UV Lambda max | 596 to 599 nm |
Absorption Ratio P-15/P+15 | 0.94 to 1.2 |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...