Tisk…

Patent blue V sodium salt

Dodavatel: Thermo Fisher Scientific

Synonyms: C.I. 42051

339330050 339330250 339331000
ACRO339330050EA 1180 CZK
ACRO339330050 ACRO339330250 ACRO339331000
Patent blue V sodium salt
Patent blue V sodium salt
Vzorec: C₂₇H₃₁N₂NaO₇S₂
MW: 582.67
Storage Temperature: Ambient
MDL Number: MFCD09608138
CAS číslo: 20262-76-4
EINECS: 222-573-8

Objednat

Výsledky specifikačního testu

Appearance Dark blue-green Crystalline powder
Infrared spectrum Conforms
Dye content ≥80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR