Eozin Y, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt
M-Clarity™ quality level = MQ200
E4382-100G
E4382-25G
SIALE4382-100GEA
0
CZK
SIALE4382-100G
SIALE4382-25G
Eozin Y, Sigma-Aldrich®
Eozin Y
Vzorec:
C₂₀H₆Br₄Na₂O₅ Bod tání: 295…296 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005040 CAS číslo: 17372-87-1 EINECS: 241-409-6 Merck Index: 13,03634 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...