Tisk…

Methyl-5-chloro-2-nitrobenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Chloro-2-nitrobenzoic acid methyl ester, Methyl 5-chloro-2-nitrobenzoate, Methyl 5-chloro-2-nitrobenzenecarboxylate

M-Clarity™ quality level = MQ100

579823-25G 579823-5G
SIAL579823-25GEA 6650 CZK
SIAL579823-25G SIAL579823-5G
Methyl-5-chloro-2-nitrobenzoate, Sigma-Aldrich®
Methyl-5-chloro-2-nitrobenzoate
Vzorec: C₈H₆ClNO₄
MW: 215,59 g/mol
Bod varu: 306 °C (1013 hPa)
Bod tání: 48,5 °C
Hustota: 1,426 g/cm³ (20 °C)
MDL Number: MFCD00272139
CAS číslo: 51282-49-6
EINECS: 257-107-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR