Tisk…

Aluminium L-trilactate, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

430633-100G 430633-500G
SIAL430633-100GEA 2250 CZK
SIAL430633-100G SIAL430633-500G
Aluminium L-trilactate, Sigma-Aldrich®
Aluminium L-trilactate
Vzorec: [CH₃CH(OH)COO]₃Al
Bod tání: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD20529175
EINECS: 242-670-9
Merck Index: 13,00347

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR