mléčnan hlinitý ≥95%
Dodavatel: abcr
|
Varování
|
Synonyms:
L-lactate aluminum, Lactic acid aluminum salt, (±)-Lactic acid aluminium salt, L-lactate aluminum salt, Aluminum lactate, (±)-2-Hydroxypropanoic acid aluminium salt, Aluminium lactate, DL-Lactic acid aluminium salt
AB119625.0100
AB119625.0500
ABCRAB119625.0100EA
0
CZK
ABCRAB119625.0100
ABCRAB119625.0500
mléčnan hlinitý ≥95%
mléčnan hlinitý
Vzorec:
(H₃CCH(OH)COO)₃Al Bod tání: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD20529175 CAS číslo: 18917-91-4 EINECS: 242-670-9 Merck Index: 13,00347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...