Tisk…

Neocuproine hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,9-Dimethyl-1,10-phenanthroline hydrate

M-Clarity™ quality level = MQ100

121908-25G 121908-5G
SIAL121908-25GEA 9490 CZK
SIAL121908-25G SIAL121908-5G
Neocuproine hydrate, Sigma-Aldrich®
Neocuproine hydrate
Vzorec: C₁₄H₁₂N₂·H₂O
MW: 226,28 g/mol
CAS číslo: 654054-57-6
EINECS: 207-601-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR