Tisk…

Neokuproin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,9-Dimethyl-1,10-phenanthroline

N1501-1G N1501-25G N1501-5G
SIALN1501-1GEA 837 CZK
SIALN1501-1G SIALN1501-25G SIALN1501-5G
Neokuproin, Sigma-Aldrich®
Neokuproin
Vzorec: C₁₄H₁₂N₂
MW: 208.26
Bod tání: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00004973
CAS číslo: 484-11-7
EINECS: 207-601-9
Merck Index: 13,06477

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR