Tisk…
ESD Safety Shoes, Lace-Up, Stewart XXG Pro GTX Low S3 HI CI
Dodavatel: ELTEN
728661/39 728661/40 728661/41 728661/42 728661/43 728661/44 728661/48 728661/45 728661/46 728661/47
ELTN728661/39EA 4080 CZK
ELTN728661/39 ELTN728661/40 ELTN728661/41 ELTN728661/42 ELTN728661/43 ELTN728661/44 ELTN728661/48 ELTN728661/45 ELTN728661/46 ELTN728661/47
ESD Safety Shoes, Lace-Up, Stewart XXG Pro GTX Low S3 HI CI
Obuv
ESD safety shoe made from hydrophobised nubuck leather upper and a rubber/PU sole (Wellmaxx Grip).
  • All weather Gore-Tex performance membrane
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Non-metallic, penetration-resistant midsole
  • TPU scuff cap
  • Composite toe cap
Colour: Black/green
Certifikace: EN ISO 20345 S3 HI/HRO/CI, form A.