Tisk…

Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PerCP (Peridinin-Chlorophyll Protein Complex))

Dodavatel: AAT Bioquest
116101T0 116101T1 116101T2
AATB116101T0EA 7230 CZK
AATB116101T0 AATB116101T1 AATB116101T2
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PerCP (Peridinin-Chlorophyll Protein Complex))
Protilátky
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Upozornení: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR