Tisk…

Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (mFluor™ Red 780)

Dodavatel: AAT BIOQUEST
101910W0 101910W1
AATB101910W0EA 9900 CZK
AATB101910W0 AATB101910W1
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (mFluor™ Red 780)
Protilátky
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Upozornení: For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: mFluor™ Red 780
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR