Tisk…
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 860)
Dodavatel: AAT BIOQUEST
100510R0 100510R1
AATB100510R0EA 9900 CZK
AATB100510R0 AATB100510R1
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 860)
Protilátky
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Upozornení: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: iFluor® 860
Epitope:
Host: Mouse
Isotype:
Reactivity: Human