Tisk…

Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 647)

Dodavatel: AAT BIOQUEST
101910F0 101910F1
AATB101910F0EA 9900 CZK
AATB101910F0 AATB101910F1
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 647)
Protilátky
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Upozornení: For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR