Tisk…

Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 633)

Dodavatel: AAT BIOQUEST
101910E0 101910E1
AATB101910E0EA 9900 CZK
AATB101910E0 AATB101910E1
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 633)
Protilátky
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Upozornení: For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 633
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu CD19 CD19
Environmentally Preferable
Název antigenu CD19 antigen CD19 antigen
Druh protilátky Primární Primární
Klonovalita Monoclonal Monoclonal
Spojení iFluor® 633 iFluor® 633
Klon SJ25C1 SJ25C1
Reaktivita Human
Hostitel Mouse Mouse
Izotyp
ImunoFluorescence Yes Yes
Průtoková cytometrie Yes Yes
Čištění Affinity chromatography Affinity chromatography
Křížová adsorbce No No
Skladovací teplota 2...8 °C with minimized light exposure. Do not freeze. 2...8 °C with minimized light exposure. Do not freeze.
Transportní teplota 20 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR