Tisk…

Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® A7)

Dodavatel: AAT BIOQUEST
103400S0 103400S1
AATB103400S0EA 9900 CZK
AATB103400S0 AATB103400S1
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® A7)
Protilátky
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Upozornení: For research use only.

Type: Primary
Antigen: CD34 antigen
Clonality: Monoclonal
Clone: 4H11
Conjugation: iFluor® A7
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR