Tisk…
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 840)
Dodavatel: AAT BIOQUEST
103400Q0 103400Q1
AATB103400Q0EA 9900 CZK
AATB103400Q0 AATB103400Q1
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 840)
Protilátky
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Upozornení: For research use only.
Type: Primary
Antigen: CD34 antigen
Clonality: Monoclonal
Clone: 4H11
Conjugation: iFluor® 840
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human