Tisk…

Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (mFluor™ Blue 570)

Dodavatel: AAT BIOQUEST INC
100520T0 100520T1
AATB100520T0EA 9900 CZK
AATB100520T0 AATB100520T1
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (mFluor™ Blue 570)
Protilátky
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Upozornení: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: mFluor™ Blue 570
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR