Tisk…

Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 430)

Dodavatel: AAT BIOQUEST
10081030 10081031
AATB10081030EA 9900 CZK
AATB10081030 AATB10081031
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 430)
Protilátky
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Upozornení: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: iFluor® 430
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR