Tisk…

Pyridinium dichromate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

214698-100G 214698-500G
SIAL214698-100GEA 2310 CZK
SIAL214698-100G SIAL214698-500G
Pyridinium dichromate, Sigma-Aldrich®
Pyridinium dichromate
Vzorec: C₁₀N₁₂Cr₂N₂O₇
Bod tání: 152…153 °C
MDL Number: MFCD00013105
CAS číslo: 20039-37-6
EINECS: 243-478-8
UN: 3087
ADR: 5.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR