Tisk…

Pyridinium dichromate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: PDC

L15132.14 L15132.22
L15132.14EA 1200 CZK
L15132.14 L15132.22
Pyridinium dichromate ≥98%
Pyridinium dichromate

Purity: 98% CAS No.: 20039-37-6 Molecular Formula: C10H12Cr2N2O7 Formula Weight: 376.21 Melting Point: ca 145-147° UN No.: 3087 MDL No.: MFCD00013105

Vzorec: C₁₀N₁₂Cr₂N₂O₇
MW: 532,19 g/mol
Bod tání: ∼ 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00013105
CAS číslo: 20039-37-6
EINECS: 243-478-8
UN: 3085
ADR: 5.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR