Tisk…

Pyridinium chlorochromate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: PCC

M-Clarity™ quality level = MQ100

190144-100G 190144-25G 190144-500G
SIAL190144-100GEA 1960 CZK
SIAL190144-100G SIAL190144-25G SIAL190144-500G
Pyridinium chlorochromate, Sigma-Aldrich®
Pyridinium chlorochromate
Vzorec: C₅H₆ClCrNO₃
Bod tání: 205 °C
MDL Number: MFCD00013106
CAS číslo: 26299-14-9
EINECS: 247-595-5
UN: 1479
ADR: 5.1,II
Merck Index: 13,08064

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR