Pyridinium chlorochromate 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
183670250
183671000
183675000
ACRO183670250EA
982
CZK
ACRO183670250
ACRO183671000
ACRO183675000
Pyridinium chlorochromate 98%
Pyridinium chlorochromate
Vzorec:
C₅H₆ClCrNO₃ Bod tání: 205 °C |
MDL Number:
MFCD00013106 CAS číslo: 26299-14-9 EINECS: 247-595-5 UN: 1479 ADR: 5.1,II Merck Index: 13,08064 |
Výsledky specifikačního testu
Appearance | Orange Crystalline powder |
Infrared spectrum | Conforms |
Titration Iodometric | 97.5 to 102.5 % |
HPLC | ≥98.5 % |
Residue after ignition | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...