Tisk…
Phenol red sodium salt, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
M-Clarity™ quality level = MQ200
114537-5G 114537-25G
SIAL114537-5GEA 1330 CZK
SIAL114537-5G SIAL114537-25G
Phenol red sodium salt, Sigma-Aldrich®
Phenol red sodium salt
Vzorec: C₁₉H₁₃NaO₅S
MW: 376,35 g/mol
Bod tání: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS číslo: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397