Phenol red sodium salt
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
B21948.06
B21948.14
B21948.A1
B21948.06EA
855
CZK
B21948.06
B21948.14
B21948.A1
Phenol red sodium salt
Phenol red sodium salt
CAS No.: 34487-61-1
Molecular Formula: C19H13NaO5S
Formula Weight: 376.37
Melting Point: 285° dec.
Beilstein Reference: 6260026
Merck Reference: 14,7247
MDL No.: MFCD00066901
Vzorec:
C₁₉H₁₃NaO₅S MW: 376.35 Bod tání: 285 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00066901 CAS číslo: 34487-61-1 EINECS: 252-057-8 Merck Index: 12,07397 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...