Tisk…

Phenol red sodium salt ACS

Dodavatel: Thermo Scientific
Varování

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

15987.06 15987.14
15987.06EA 1200 CZK
15987.06 15987.14
Phenol red sodium salt ACS
Phenol red sodium salt

CAS No.: 34487-61-1 Molecular Formula: C19H13NaO5S Formula Weight: 376.37 Physical Form: Crystalline Melting Point: 285° dec. Beilstein Reference: 6260026 Merck Reference: 14,7247 MDL No.: MFCD00066901


pH indicator

Vzorec: C₁₉H₁₃NaO₅S
MW: 376,35 g/mol
Bod tání: 285 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS číslo: 34487-61-1
EINECS: 252-057-8
Merck Index: 14,7247

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR