Tisk…

Disodium oxide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium oxide

M-Clarity™ quality level = MQ100

645613-250G 645613-50G
SIAL645613-250GEA 10670 CZK
SIAL645613-250G SIAL645613-50G
Disodium oxide, Sigma-Aldrich®
Disodium oxide
Vzorec: Na₂O
MDL Number: MFCD00046201
CAS číslo: 1313-59-3
EINECS: 215-208-9
UN: 1825
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR