Lauroyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Dodecanoyl chloride
M-Clarity™ quality level = MQ200
156930-1L
156930-250ML
156930-50ML
SIAL156930-1LEA
5210
CZK
SIAL156930-1L
SIAL156930-250ML
SIAL156930-50ML
Lauroyl chloride, Sigma-Aldrich®
Lauroyl chloride
Vzorec:
C₁₂H₂₃ClO MW: 218.77 Bod varu: 267 Bod tání: –12 °C Hustota: 0.922 |
MDL Number:
MFCD00000740 CAS číslo: 112-16-3 EINECS: 203-941-7 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...