Lauroyl chloride ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13294.22
A13294.36
A13294.22EA
1160
CZK
A13294.22
A13294.36
Lauroyl chloride ≥98%
Lauroyl chloride
Purity: 98%
CAS No.: 112-16-3
Molecular Formula: C12H23ClO
Formula Weight: 218.77
Flash Point: 140°(284°F)
Refractive Index: 1.4460
Melting Point: -17°
Boiling Point: 130-132°/7mm
Beilstein Reference: 1281201
UN No.: 3265
MDL No.: MFCD00000740
Vzorec:
C₁₂H₂₃ClO Bod varu: 130…132 °C (7 mmHg) Bod tání: –17 °C Hustota: 0,922 Bod vzplanutí: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000740 CAS číslo: 112-16-3 EINECS: 203-941-7 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...