Decanoyl chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
140295-100ML
140295-500ML
SIAL140295-100MLEA
1620
CZK
SIAL140295-100ML
SIAL140295-500ML
Decanoyl chloride, Sigma-Aldrich®
Decanoyl chloride
Vzorec:
C₁₀H₁₉ClO Bod varu: 233 °C (1013 hPa) Hustota: 0,919 g/cm³ (20 °C) |
MDL Number:
MFCD00000771 CAS číslo: 112-13-0 EINECS: 203-938-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...