Tisk…

Bentonit, grey powder

Dodavatel: MP Biomedicals

Synonyms: Montmorillonite, Bentonite

0210089410 02100894.5
ICNA0210089410EA 21240 CZK
ICNA0210089410 ICNA02100894.5
Bentonit, grey powder
Bentonite

Bentonite is used as aluminum silicate (clay), emulsifier and adsorbent for proteins and virus particles.

Hustota: 2,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00130611
CAS číslo: 1302-78-9
EINECS: 215-108-5

Objednat

Výsledky specifikačního testu

Identity Test Passes
pH (2% aq soln) 9,5 - 10,5
Loss on Drying (2 hr) ≤12%
Viscosity (2% aq solution) 8 - 30 cps

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR