Tisk…

Triamterene 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2,4,7-Triamino-6-phenylpteridine, 6-Phenyl-2,4,7-triaminopteridine

B20044.06 B20044.14 B20044.22
B20044.06EA 1410 CZK
B20044.06 B20044.14 B20044.22
Triamterene 98%
Triamterene

Purity: 98% CAS No.: 396-01-0 Molecular Formula: C12H11N7 Formula Weight: 253.27 Melting Point: 300° Beilstein Reference: 266723 Merck Reference: 14,9599 MDL No.: MFCD00006708

Vzorec: C₁₂H₁₁N₇
MW: 253,27 g/mol
Bod varu: 573 °C (1013 hPa)
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006708
CAS číslo: 396-01-0
EINECS: 206-904-3
Merck Index: 14,9599

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR