5-Chloro-2-thiopheneboronic acid 97+%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(5-Chlorothiophen-2-yl)boronic acid, 5-Chloro-2-thienylboronic acid, 5-Chlorothiophene-2-boronic acid
359010010
359010050
ACRO359010010EA
956
CZK
ACRO359010010
ACRO359010050
5-Chloro-2-thiopheneboronic acid 97+%
5-Chloro-2-thiopheneboronic acid
Vzorec:
C₄H₄BClO₂S MW: 162,4 g/mol Bod tání: 136…140 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00236030 CAS číslo: 162607-18-3 |
Výsledky specifikačního testu
Appearance | Off-white to light green to light brown Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Titration with NaOH | 97.0 to 103.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...