Tisk…

Bromocresol purple sodium salt

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Sodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonate, Bromocresol purple water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolate

A19082.09 A19082.18
A19082.09EA 1390 CZK
A19082.09 A19082.18
Bromocresol purple sodium salt
Bromocresol purple sodium salt

CAS No.: 62625-30-3 Molecular Formula: C21H15Br2NaO5S Formula Weight: 562.22 Melting Point: ca 255° dec. MDL No.: MFCD00148896

Vzorec: C₂₁H₁₅Br₂NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00148896
CAS číslo: 62625-30-3
EINECS: 263-655-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR