Tisk…

4-(Dimethylamino)benzaldehyd, crystalline powder ACS

Dodavatel: MP Biomedicals
Varování

0215251783 0215251750
ICNA0215251783EA 7970 CZK
ICNA0215251783 ICNA0215251750
4-(Dimethylamino)benzaldehyd, crystalline powder ACS
4-(Dimethylamino)benzaldehyde

Soluble in alcohol (40 mg/ml - ethanol), HCl (50 mg/ml - 10% HCl), benzene, chloroform, ether; slightly soluble in water.


  • UV/Visible Absorbance : λ min 369 nm; λ max 385 nm(Lit.)
  • Presentation : Off-white Crystalline Powder
  • Color (APHA) : ≤60 of Alcohol Solution
  • Residue on ignition : ≤0.1%
  • Solubility : Soluble in alcohol (40 mg/mL-ethanol), HCl (50 mg/mL-10% HCl), benzene, chloroform, ether; slightly soluble in water.


Storage: protect from light, under nitrogen.

Vzorec: (CH₃)₂NC₆H₄CHO
Bod varu: 176,..177 °C (17 mmHg) (lit.)
Hustota: 1,0254 g/cm³ (99.9 °C)
Bod vzplanutí: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS číslo: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Identity Test Passes
Melting Point 73 - 75°C
Solubility in Alcohol Passes
Color (APHA) of Alcohol
Solution
<60
Solubility in Hydrochloric
Acid
Passes
Color of Hydrochloric Acid
Solution
Passes
Residue after Ignition <0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR