Tisk…
Patton-reeders reagent (Calconcarboxylic acid dihydrate), Sigma-Aldrich®
Dodavatel: Merck
Varování
Synonyms: 2,2'-Dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid dihydrate
M-Clarity™ quality level = MQ100
H3128-100G H3128-25G H3128-5G
SIALH3128-100GEA 30120 CZK
SIALH3128-100G SIALH3128-25G SIALH3128-5G
Patton-reeders reagent (Calconcarboxylic acid dihydrate), Sigma-Aldrich®
Patton-reeders reagent (Calconcarboxylic acid dihydrate)
Vzorec: C₂₁H₁₄N₂O₇S·2H₂O
MW: 474,45 g/mol
Bod tání: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004078
CAS číslo: 3737-95-9
EINECS: 223-117-0