3-Thiophenecarbonitrile ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Cyanothiophene, 3-Thienyl cyanide, Thiophene-3-carbonitrile
L13115.06
L13115.14
L13115.06EA
1880
CZK
L13115.06
L13115.14
3-Thiophenecarbonitrile ≥98%
3-Thiophenecarbonitrile
Purity: 98%
CAS No.: 1641-09-4
Molecular Formula: C5H3NS
Formula Weight: 109.15
Flash Point: 86°(186°F)
Refractive Index: 1.5615
Odour: Recognizable
Boiling Point: 205°
Beilstein Reference: 107104
UN No.: 3276
MDL No.: MFCD00151852
Vzorec:
C₅H₃NS MW: 109,15 g/mol Bod varu: 205 °C Hustota: 1,200 Bod vzplanutí: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00151852 CAS číslo: 1641-09-4 EINECS: 216-687-7 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...