2-Thiopheneboronic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Thiophen-2-ylboronic acid, Thiophene-2-boronic acid, 2-Thienylboronic acid
M-Clarity™ quality level = MQ200
436836-1G
436836-5G
SIAL436836-1GEA
769
CZK
SIAL436836-1G
SIAL436836-5G
2-Thiopheneboronic acid, Sigma-Aldrich®
2-Thiopheneboronic acid
Vzorec:
C₄H₅BO₂S MW: 127,96 g/mol Bod tání: 146…147 °C |
MDL Number:
MFCD00151850 CAS číslo: 6165-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...