Tisk…

2-Thiophenecarbonitrile ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Thienyl cyanide, Thiophene-2-carbonitrile, 2-Cyanothiophene

A11930.14 A11930.22 A11930.36
A11930.14EA 3380 CZK
A11930.14 A11930.22 A11930.36
2-Thiophenecarbonitrile ≥98%
2-Thiophenecarbonitrile

Purity: 98% CAS No.: 1003-31-2 Molecular Formula: C5H3NS Formula Weight: 109.15 Flash Point: 81°(178°F) Refractive Index: 1.5645 Odour: Unpleasant Boiling Point: 191-193° Beilstein Reference: 107035 UN No.: 3276 MDL No.: MFCD00005416

Vzorec: C₅H₃NS
MW: 109,15 g/mol
Bod varu: 191…193 °C
Hustota: 1,171
Bod vzplanutí: 81 °C (178 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005416
CAS číslo: 1003-31-2
EINECS: 213-706-0
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR