Methyl red hydrochloride ACS
Dodavatel: Thermo Scientific
Synonyms:
2-(4-Dimethylaminophenylazo)benzoic acid hydrochloride
339320250
339321000
ACRO339320250EA
1170
CZK
ACRO339320250
ACRO339321000
Methyl red hydrochloride ACS
Methyl red hydrochloride
Vzorec:
C₁₅H₁₆ClN₃O₂ MW: 305,76 g/mol Bod tání: 175 °C |
MDL Number:
MFCD00012614 CAS číslo: 63451-28-5 EINECS: 264-190-9 |
Výsledky specifikačního testu
Appearance | Blue to dark purple Powder |
Infrared spectrum | Conforms |
Visual transition interval | From pH 4.2 (pink) to pH 6.2 (yellow) |
Clarity of alcohol solution | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...