3-Chlorostyrene ≥98% stabilised
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
m-Chlorostyrene
Stabilization:
0.10 % Stabilised with 4-tert-butylpyrocatechol (TBC)
B23806.06
B23806.14
B23806.22
B23806.06EA
2590
CZK
B23806.06
B23806.14
B23806.22
3-Chlorostyrene ≥98% stabilised
3-Chlorostyrene
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 98%
CAS No.: 2039-85-2
Molecular Formula: C8H7Cl
Formula Weight: 138.60
Storage Temperature: 2°C to 8°C
Flash Point: 67°(153°F)
Refractive Index: 1.5620
Odour: Characteristic
Boiling Point: 188-189°
Beilstein Reference: 2038489
MDL No.: MFCD00000598
Vzorec:
C₈H₇Cl MW: 138.6 Bod varu: 188…189 °C Bod tání: –26 °C Hustota: 1.090 Bod vzplanutí: 67 °C (153 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000598 CAS číslo: 2039-85-2 EINECS: 218-024-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...