4-Bromostyrene ≥98% stabilizovaný
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Bromo-4-vinylbenzene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0,10 %
A17896.06
A17896.14
A17896.22
A17896.06EA
1960
CZK
A17896.06
A17896.14
A17896.22
4-Bromostyrene ≥98% stabilizovaný
4-Bromostyrene
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 98%
CAS No.: 2039-82-9
Molecular Formula: C8H7Br
Formula Weight: 183.05
Storage Temperature: 2°C to 8°C
Flash Point: 75°(167°F)
Refractive Index: 1.5940
Melting Point: 3-6°
Boiling Point: 88-89°/16mm
Beilstein Reference: 1634204
MDL No.: MFCD00000110
Vzorec:
C₈H₇Br MW: 183,05 g/mol Bod varu: 88…89 °C (16 mmHg) Bod tání: 3…6 °C Hustota: 1,400 Bod vzplanutí: 75 °C (167 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000110 CAS číslo: 2039-82-9 EINECS: 218-022-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...