Tisk…

4-Bromostyrene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Bromo-4-vinylbenzene

M-Clarity™ quality level = MQ200

124141-10G 124141-25G
SIAL124141-10GEA 4970 CZK
SIAL124141-10G SIAL124141-25G
4-Bromostyrene, Sigma-Aldrich®
4-Bromostyrene
Vzorec: C₈H₇Br
Bod varu: 214 °C (1013 hPa)
Hustota: 1,5991 g/cm³ (20 °C)
MDL Number: MFCD00000110
CAS číslo: 2039-82-9
EINECS: 218-022-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR