Tisk…

2-Bromostyrene 96% stabilizovaný

Dodavatel: Thermo Scientific
Varování


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC) 0,05 %

B24181.06 B24181.14
B24181.06EA 4530 CZK
B24181.06 B24181.14
2-Bromostyrene 96% stabilizovaný
2-Bromostyrene

Stabilized with ca 0.05% 4-tert-butylcatechol


Purity: 97% CAS No.: 2039-88-5 Molecular Formula: C8H7Br Formula Weight: 183.05 Storage Temperature: 2°C to 8°C Flash Point: 85°(185°F) Refractive Index: 1.5920 Melting Point: -53° Boiling Point: 102-104°/22mm Beilstein Reference: 2323537 MDL No.: MFCD00000076

Vzorec: C₈H₇Br
MW: 183,05 g/mol
Bod varu: 102…104 °C (22 mmHg)
Bod tání: –53 °C
Hustota: 1,416
Bod vzplanutí: 85 °C (185 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000076
CAS číslo: 2039-88-5
EINECS: 218-027-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR