2,5-Dimethylstyrene ≥97% stabilised
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
B21476.03
B21476.06
B21476.14
B21476.03EA
1180
CZK
B21476.03
B21476.06
B21476.14
2,5-Dimethylstyrene ≥97% stabilised
2,5-Dimethylstyrene
Stabilized with 4-tert-butylcatechol
Purity: 97%
CAS No.: 2039-89-6
Molecular Formula: C10H12
Formula Weight: 132.21
Storage Temperature: 2°C to 8°C
Flash Point: 63°(145°F)
Refractive Index: 1.5390
Melting Point: -35°
Boiling Point: 83-84°/24mm
Beilstein Reference: 2203656
MDL No.: MFCD00008614
Vzorec:
C₁₀H₁₂ Bod varu: 83…84 °C (24 mmHg) Bod tání: –35 °C Hustota: 0,904 Bod vzplanutí: 63 °C (145 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008614 CAS číslo: 2039-89-6 EINECS: 218-028-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...