Tisk…

4-Vinylveratrole, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,4-Dimethoxystyrene, 1,2-Dimethoxy-4-vinylbenzene

M-Clarity™ quality level = MQ100

154466-10G 154466-5G
SIAL154466-10GEA 9470 CZK
SIAL154466-10G SIAL154466-5G
4-Vinylveratrole, Sigma-Aldrich®
4-Vinylveratrole
Vzorec: C₁₀H₁₂O₂
MW: 164.2
Storage Temperature: Refrigerator
MDL Number: MFCD00008615
CAS číslo: 6380-23-0
EINECS: 228-962-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR