Ethylendi(acetát), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Ethane-1,2-diyl diacetate, Ethylene glycol diacetate, Ethylene Glycol Diacetate, 1,2-Diacetoxyethane
M-Clarity™ quality level = MQ200
525200-20L
525200-4L
525200-500ML
SIAL525200-20LEA
18310
CZK
SIAL525200-20L
SIAL525200-4L
SIAL525200-500ML
Ethylendi(acetát), Sigma-Aldrich®
Ethylendi(acetát)
Vzorec:
C₆H₁₀O₄ MW: 146.14 Bod varu: 190…191 °C (1013 hPa) Bod tání: –31 °C Hustota: 1.104 g/cm³ (20 °C) Bod vzplanutí: 88 Storage Temperature: Ambient |
MDL Number:
MFCD00008718 CAS číslo: 111-55-7 EINECS: 203-881-1 Merck Index: 13,03833 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...