Tisk…
EDTA železitá monosodná sůl, Sigma-Aldrich®
Dodavatel: Merck
Varování
Synonyms: Ethylenedinitrilotetra-acetic acid iron (III) sodium salt, Edetic acid iron (III) monosodium salt, Edetic acid iron (III) sodium salt, Ethylenediaminetetra-acetic acid iron (III) sodium salt, Iron (III) sodium ethylenediaminetetraacetate, Ethylenedinitrilotetra-acetic acid iron (III) monosodium salt, EDTA iron sodium, Sodium ferric ethylenediaminetetraacetate, Iron (III) monosodium ethylenediaminetetraacetate, Sodium feredetate, Ethylenediaminetetra-acetic acid iron (III) monosodium salt
MQ200
E6760-100G E6760-500G
SIALE6760-100GEA 2740 CZK
SIALE6760-100G SIALE6760-500G
EDTA železitá monosodná sůl, Sigma-Aldrich®
EDTA železitá monosodná sůl
Vzorec: C₁₀H₁₂FeN₂NaO₈
MW: 367.05
Bod tání: 80
Storage Temperature: Ambient
MDL Number: MFCD00078215
CAS číslo: 15708-41-5
EINECS: 239-802-2
Merck Index: 12,04076