Tisk…

EDTA železitá monosodná sůl

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ethylenedinitrilotetra-acetic acid iron (III) sodium salt, Edetic acid iron (III) monosodium salt, Edetic acid iron (III) sodium salt, Ethylenediaminetetra-acetic acid iron (III) sodium salt, Iron (III) sodium ethylenediaminetetraacetate, Ethylenedinitrilotetra-acetic acid iron (III) monosodium salt, EDTA iron sodium, Sodium ferric ethylenediaminetetraacetate, Iron (III) monosodium ethylenediaminetetraacetate, Sodium feredetate, Ethylenediaminetetra-acetic acid iron (III) monosodium salt

88995.22 88995.36 88995.A3
88995.22EA 1490 CZK
88995.22 88995.36 88995.A3
EDTA železitá monosodná sůl
EDTA železitá monosodná sůl

CAS No.: 15708-41-5 Molecular Formula: FeNa(O2CCH2)2NCH2CH2N(CH2CO2)2 Formula Weight: 367.05 Physical Form: Powder Odour: Odourless MDL No.: MFCD00078215

Vzorec: C₁₀H₁₂FeN₂NaO₈
MW: 367.05
Bod tání: 80
Storage Temperature: Ambient
MDL Number: MFCD00078215
CAS číslo: 15708-41-5
EINECS: 239-802-2
Merck Index: 12,04076

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR