Tisk…
Anti-IgG Donkey Polyclonal Antibody (CF® 583R)
Dodavatel: Biotium
20897-500UL 20897-50UL
20897-500ULEA 7050 CZK
20897-500UL 20897-50UL
Anti-IgG Donkey Polyclonal Antibody (CF® 583R)
Protilátky
This is a highly cross-adsorbed donkey anti-sheep IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimize cross-reactivity, the antibody has been adsorbed against chicken, guinea pig, horse, human, mouse, rabbit, rat, and Syrian hamster.
  • Highly cross-adsorbed for specific staining with minimal background
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
Concentration: 2 mg/ml
Antibody/conjugate formulation-Liquid: PBS/50% glycerol/2 mg/ml BSA/0,05% azide, Lyophilised: PBS/15 mg/ml BSA/20 mg/ml trehalose after reconstitution
Secondary/tag antibody applications: Flow cytometry, IHC, IF (cells or tissue sections), western blot